Learn More
Cayman Chemical 1Oleoyl2hydroxysnglycero 10mg
SDP
Catalog No. 502981741
Encompass_Preferred
A lysophospholipid containing oleic acid (18:1) at the sn-1 position
Catalog No. 50-298-1741 Supplier Cayman Chemical Supplier No. 2096010 MG
A lysophospholipid containing oleic acid (18:1) at the sn-1 position