Learn More
Cayman Chemical 1-Oleoyl-2-hydroxy-sn-glycero-3-phosphocholine, Purity: ≥95%, CAS: 19420-56-5, 10 mg
SDP
Catalog No. 502983050
Encompass_Preferred
A lysophospholipid; inhibits taurine uptake in Caco-2 cells at 200 µM; conditioned medium levels are increased in VEGF-stimulated primary NHBE cells, an effect that can be reversed by oleyloxyethyl phosphorylcholine; plasma levels are decreased in patients with lung cancer or abdominal aortic aneurysm
Catalog No. 50-298-3050 Supplier Cayman Chemical Supplier No. 2095910 MG
A lysophospholipid; inhibits taurine uptake in Caco-2 cells at 200 µM; conditioned medium levels are increased in VEGF-stimulated primary NHBE cells, an effect that can be reversed by oleyloxyethyl phosphorylcholine; plasma levels are decreased in patients with lung cancer or abdominal aortic aneurysm