Learn More
Cayman Chemical 1-Oleoyl-2-hydroxy-sn-glycero-3-PE, Purity: ≥95%, CAS: 89576-29-4, 50 mg
SDP
Catalog No. 502936554
Encompass_Preferred
A naturally-occurring lysophospholipid; induces transient increases in intracellular calcium in PC12 rat neuronal cells in a concentration-dependent manner, an effect which can be blocked by the LPA1 antagonist AM095; increases IL-2 production in CD1d-stimulated murine NKT cell hybridoma 2H4 cells at 100 ng/ml; induces production of IL-4, but not IFN-γ, in murine splenocytes at 100 ng/ml and in mouse serum at 1 μg per animal; levels increase in iBAT in mice following three-day cold exposure
Catalog No. 50-293-6554 Supplier Cayman Chemical Supplier No. 2559650 MG
A naturally-occurring lysophospholipid; induces transient increases in intracellular calcium in PC12 rat neuronal cells in a concentration-dependent manner, an effect which can be blocked by the LPA1 antagonist AM095; increases IL-2 production in CD1d-stimulated murine NKT cell hybridoma 2H4 cells at 100 ng/ml; induces production of IL-4, but not IFN-γ, in murine splenocytes at 100 ng/ml and in mouse serum at 1 μg per animal; levels increase in iBAT in mice following three-day cold exposure