Learn More
Cayman Chemical 1OOctadecyl2Omethylsng 100mg
SDP
Catalog No. 502970143
Encompass_Preferred
A metabolite of a phosphotidylinositol ether lipid analog known to target the pleckstrin homology domain of Akt and to induce apoptosis in cancer cell lines with high levels of endogenous Akt activity
Catalog No. 50-297-0143 Supplier Cayman Chemical Supplier No. 15108100 MG
A metabolite of a phosphotidylinositol ether lipid analog known to target the pleckstrin homology domain of Akt and to induce apoptosis in cancer cell lines with high levels of endogenous Akt activity