Learn More
Cayman Chemical 1OOctadecyl2Omethylsng 50mg
SDP
Catalog No. 502973230
Encompass_Preferred
A metabolite of a phosphotidylinositol ether lipid analog known to target the pleckstrin homology domain of Akt and to induce apoptosis in cancer cell lines with high levels of endogenous Akt activity
Catalog No. 50-297-3230 Supplier Cayman Chemical Supplier No. 1510850 MG
A metabolite of a phosphotidylinositol ether lipid analog known to target the pleckstrin homology domain of Akt and to induce apoptosis in cancer cell lines with high levels of endogenous Akt activity