Learn More
Selleck Chemical LLC IOX1 10mg 5852-78-8
SDP
Supplier:  Selleck Chemical LLC S723410MG
Encompass_Preferred
IOX1 is a potent and broad-spectrum inhibitor of 2OG oxygenases, including the JmjC demethylases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1001456