Learn More
Cayman Chemical 1-Oxyl-2,2,5,5-tetramethylpyrroline-3-carboxylate NHS ester, Purity: ≥98%, CAS: 37558-29-5, 50 mg
SDP
Catalog No. 502970431
Encompass_Preferred
An N-hydroxysuccinimide ester of the nitroxide spin label tempyo; an active acylating agent that preferentially targets α-amino groups, used to spin label proteins and aminoacylated RNA species
Catalog No. 50-297-0431 Supplier Cayman Chemical Supplier No. 1614850 MG
An N-hydroxysuccinimide ester of the nitroxide spin label tempyo; an active acylating agent that preferentially targets α-amino groups, used to spin label proteins and aminoacylated RNA species