Learn More
Cayman Chemical 1P-ETH-LAdolutIon 100ug
SDP
Catalog No. 502943776
Encompass_Preferred
An analytical reference standard categorized as a lysergamide; intended for research and forensic applications
Catalog No. 50-294-3776 Supplier Cayman Chemical Supplier No. 39805100 UG
An analytical reference standard categorized as a lysergamide; intended for research and forensic applications