Learn More
Cayman Chemical 1PalmItol2Docosahexaenol 10mg
SDP
Catalog No. 502947469
Encompass_Preferred
A phospholipid; a component of LDL and HDL; enrichment in recombinant HDLs decreases cholesterol ester formation by LCAT in vitro
Catalog No. 50-294-7469 Supplier Cayman Chemical Supplier No. 3198210 MG
A phospholipid; a component of LDL and HDL; enrichment in recombinant HDLs decreases cholesterol ester formation by LCAT in vitro