Learn More
Cayman Chemical 1PalmItoyl2 17 18EpETE 100ug
SDP
Catalog No. 502936211
Encompass_Preferred
An ω-3 epoxidated eicosapentaenoic acid- and palmitic acid-containing form of phosphatidylcholine; hydrolyzed by PAF-AH2 to release 17(18)-EpETE
Catalog No. 50-293-6211 Supplier Cayman Chemical Supplier No. 24383100 UG
An ω-3 epoxidated eicosapentaenoic acid- and palmitic acid-containing form of phosphatidylcholine; hydrolyzed by PAF-AH2 to release 17(18)-EpETE