Learn More
Cayman Chemical 1PalmItoyl213 SHODEsng 100ug
SDP
Catalog No. 502942674
Encompass_Preferred
An oxidized phospholipid; has been used to detect GPX4 activity using 1-palmitoyl-2-13(S)-HpODE-sn-glycero-3-PC as the substrate
Catalog No. 50-294-2674 Supplier Cayman Chemical Supplier No. 41445100 UG
An oxidized phospholipid; has been used to detect GPX4 activity using 1-palmitoyl-2-13(S)-HpODE-sn-glycero-3-PC as the substrate