Learn More
Cayman Chemical 1-Palmitoyl-2-Arachidonoyl-sn-glycero-3-PC, Purity: ≥95%, CAS: 35418-58-7, 100 mg
SDP
Catalog No. 502936378
Encompass_Preferred
A phospholipid; found in biological membranes; is oxidized in vivo and has been used to study signaling of oxidized phospholipids; levels are decreased in isolated human multiple myeloma cells
Catalog No. 50-293-6378 Supplier Cayman Chemical Supplier No. 25658100 MG
A phospholipid; found in biological membranes; is oxidized in vivo and has been used to study signaling of oxidized phospholipids; levels are decreased in isolated human multiple myeloma cells