Learn More
Cayman Chemical 1-Palmitoyl-2-Docosahexaenoyl-sn-glycero-3-PE, Purity: ≥95%, CAS: 96998-00-4, 5 mg
SDP
Catalog No. 502947336
Encompass_Preferred
A phospholipid; found in the nucleus, but not the mitochondria, of mouse liver; levels are lower in pre-term infant plasma compared with maternal and fetal plasma
Catalog No. 50-294-7336 Supplier Cayman Chemical Supplier No. 334405 MG
A phospholipid; found in the nucleus, but not the mitochondria, of mouse liver; levels are lower in pre-term infant plasma compared with maternal and fetal plasma