Learn More
Cayman Chemical 1-Palmitoyl-2-hydroxy-sn-glycero-3-PE, Purity: ≥95% (9:1 mixture of 1-acyl and 2-acyl), CAS: 53862-35-4, 10 mg
SDP
Catalog No. 502936813
Encompass_Preferred
A lysophospholipid; inhibits the growth of L. donovani promastigotes (GIC50 = 8 µM); serum levels are decreased in a mouse model of alcohol-induced liver injury and in a hepatocellular carcinoma mouse xenograft model; human serum levels are decreased immediately and 14 hours following an exercise regimen of 2.5 hours of running for three days; has been used as an internal standard for the quantification of saturated lysophosphoethanolamines
Catalog No. 50-293-6813 Supplier Cayman Chemical Supplier No. 2601110 MG
A lysophospholipid; inhibits the growth of L. donovani promastigotes (GIC50 = 8 µM); serum levels are decreased in a mouse model of alcohol-induced liver injury and in a hepatocellular carcinoma mouse xenograft model; human serum levels are decreased immediately and 14 hours following an exercise regimen of 2.5 hours of running for three days; has been used as an internal standard for the quantification of saturated lysophosphoethanolamines