Learn More
Cayman Chemical 1-Palmitoyl-2-Lauroyl-sn-glycero-3-PC, Purity: ≥95%, CAS: 82765-47-7, 100 mg
SDP
Catalog No. 502970085
Encompass_Preferred
A phospholipid in which the two acyl substituents at positions sn-1 and sn-2 are specified as palmitoyl (16:0) and lauroyl (12:0), respectively
Catalog No. 50-297-0085 Supplier Cayman Chemical Supplier No. 15101100 MG
A phospholipid in which the two acyl substituents at positions sn-1 and sn-2 are specified as palmitoyl (16:0) and lauroyl (12:0), respectively