Learn More
Cayman Chemical 1PalmItoyl2LInoleoylsngl 10mg
SDP
Supplier:  Cayman Chemical 2096210 MG
Encompass_Preferred
A phospholipid containing palmitic (16:0) and linoleic (18:2) acids inserted at the sn-1 and sn-2 positions, respectively
Catalog No. 50-298-3052