Learn More
Cayman Chemical 1PalmItoyl2oleoyl3lInole 100mg
SDP
Catalog No. 502894133
Encompass_Preferred
POL is a common triglyceride component in seed and vegetable oils including olive, sesame, soybean, canola, corn, hazelnut, and many others. POL is one of the standard triglyceride components used for the USP analysis of sesame oil for pharmaceutical applications.{8754}
Catalog No. 50-289-4133 Supplier Cayman Chemical Supplier No. 62660100 MG
POL is a common triglyceride component in seed and vegetable oils including olive, sesame, soybean, canola, corn, hazelnut, and many others. POL is one of the standard triglyceride components used for the USP analysis of sesame oil for pharmaceutical applications.{8754}