Learn More
Cayman Chemical 1-Palmitoyl-2-Oleoyl-3-Linoleoyl-rac-glycerol, Purity: ≥85%, CAS: 2680-59-3, 50 mg
SDP
Catalog No. 502892499
Encompass_Preferred
POL is a common triglyceride component in seed and vegetable oils including olive, sesame, soybean, canola, corn, hazelnut, and many others. POL is one of the standard triglyceride components used for the USP analysis of sesame oil for pharmaceutical applications.{8754}
Catalog No. 50-289-2499 Supplier Cayman Chemical Supplier No. 6266050 MG
POL is a common triglyceride component in seed and vegetable oils including olive, sesame, soybean, canola, corn, hazelnut, and many others. POL is one of the standard triglyceride components used for the USP analysis of sesame oil for pharmaceutical applications.{8754}