Learn More
Cayman Chemical 1-Palmitoyl-2-Oleoyl-3-Stearoyl-rac-glycerol, Purity: ≥98%, CAS: 2190-27-4, 5 mg
SDP
Catalog No. 502938107
Encompass_Preferred
A triacylglycerol; one of the primary triacylglycerols found in cocoa butter
Catalog No. 50-293-8107 Supplier Cayman Chemical Supplier No. 270675 MG
A triacylglycerol; one of the primary triacylglycerols found in cocoa butter