Learn More
Cayman Chemical 1PalmItoyl2Oleoylracglyc 10mg
SDP
Supplier:  Cayman Chemical 2716410 MG
Encompass_Preferred
A diacylglycerol; has been found in crude sunflower lecithin; levels increase in RINm5F rat insulinoma cells treated with oleate, elaidate, or trans-vaccenate
Catalog No. 50-293-9562