Learn More
Cayman Chemical 1-Palmitoyl-2-Oleoyl-sn-glycero-3-PG (sodium salt), Purity: ≥98%, CAS: 268550-95-4, 100 mg
SDP
Catalog No. 502970102
Encompass_Preferred
A phospholipid containing palimitoyl (16:0) and oleoyl (18:1) acyl substituents at the sn-1 and sn-2 positions, respectively, that been used to generate lipid membrane bilayers with controlled permeability and may be useful for various surfactant applications
Catalog No. 50-297-0102 Supplier Cayman Chemical Supplier No. 15105100 MG
A phospholipid containing palimitoyl (16:0) and oleoyl (18:1) acyl substituents at the sn-1 and sn-2 positions, respectively, that been used to generate lipid membrane bilayers with controlled permeability and may be useful for various surfactant applications