Learn More
Cayman Chemical 1PalmItoyl2Oleoylsnglyce 100mg
SDP
Supplier:  Cayman Chemical 20067100 MG
Encompass_Preferred
A phospholipid; a major component of E. coli bilayers; vesicles composed of 1,2,-POPE can be disrupted by polycationic antimicrobials; has been used in combination with other lipids in the formation of LNPs for spleen- and liver-specific targeting in mice
Catalog No. 50-298-4833