Learn More
Cayman Chemical 1-Palmitoyl-2-Oleoyl-sn-glycero-3-PC, Purity: ≥95%, CAS: 26853-31-6, 100 mg
SDP
Catalog No. 502970100
Encompass_Preferred
Phospholipids used for liposome production in order to study the properties of lipid bilayers
Catalog No. 50-297-0100 Supplier Cayman Chemical Supplier No. 15102100 MG
Phospholipids used for liposome production in order to study the properties of lipid bilayers