Learn More
Cayman Chemical 1PalmItoyl2Oleoylsnglyce 50mg
SDP
Catalog No. 502973228
Encompass_Preferred
A phospholipid containing palimitoyl (16:0) and oleoyl (18:1) acyl substituents at the sn-1 and sn-2 positions, respectively, that been used to generate lipid membrane bilayers with controlled permeability and may be useful for various surfactant applications
Catalog No. 50-297-3228 Supplier Cayman Chemical Supplier No. 1510550 MG
A phospholipid containing palimitoyl (16:0) and oleoyl (18:1) acyl substituents at the sn-1 and sn-2 positions, respectively, that been used to generate lipid membrane bilayers with controlled permeability and may be useful for various surfactant applications