Learn More
Cayman Chemical 1PalmItoyl2PyrpheophorbId 10mg
SDP
Catalog No. 502917877
Encompass_Preferred
A phospholipid-porphyrin conjugate; LNPs containing 1-palmitoyl-2-pyropheophorbide a-sn-glycero-3-PC and encapsulating FAM-labeled siRNA increase cytosolic siRNA levels; 1-palmitoyl-2-pyropheophorbide a-sn-glycero-3-PC-stabilized nanoemulsions loaded with paclitaxel and combined with laser irradiation reduce tumor growth and increase survival in a KB squamous cell carcinoma mouse xenograft model
Catalog No. 50-291-7877 Supplier Cayman Chemical Supplier No. 3843010 MG
A phospholipid-porphyrin conjugate; LNPs containing 1-palmitoyl-2-pyropheophorbide a-sn-glycero-3-PC and encapsulating FAM-labeled siRNA increase cytosolic siRNA levels; 1-palmitoyl-2-pyropheophorbide a-sn-glycero-3-PC-stabilized nanoemulsions loaded with paclitaxel and combined with laser irradiation reduce tumor growth and increase survival in a KB squamous cell carcinoma mouse xenograft model