Learn More
Cayman Chemical 1PalmItoyl2Stearoyl3Buty 10mg
SDP
Catalog No. 502934767
Encompass_Preferred
A triacylglycerol; has been used to determine the stereospecificity and typoselectivity of lipases
Catalog No. 50-293-4767 Supplier Cayman Chemical Supplier No. 2684210 MG
A triacylglycerol; has been used to determine the stereospecificity and typoselectivity of lipases