Learn More
Cayman Chemical 1PalmItoyl2Stearoyl3Oleo 10mg
SDP
Catalog No. 502937433
Encompass_Preferred
A triacylglycerol; has been detected in RAW 264.7 cells by neutral loss MS; increased serum levels are a potential biomarker for NAFLD
Catalog No. 50-293-7433 Supplier Cayman Chemical Supplier No. 2706810 MG
A triacylglycerol; has been detected in RAW 264.7 cells by neutral loss MS; increased serum levels are a potential biomarker for NAFLD