Learn More
Cayman Chemical 1-Palmitoyl-2-Stearoyl-rac-glycerol, Purity: ≥95%, CAS: 20296-26-8, 10 mg
SDP
Catalog No. 502932914
Encompass_Preferred
A diacylglycerol; has been used for the determination of the diacylglycerol fatty acyl chain properties that activate ARF1 GAP
Catalog No. 50-293-2914 Supplier Cayman Chemical Supplier No. 2688410 MG
A diacylglycerol; has been used for the determination of the diacylglycerol fatty acyl chain properties that activate ARF1 GAP