Learn More
Cayman Chemical 1PalmItoyl3LInolenoylrac 10mg
SDP
Catalog No. 502934200
Encompass_Preferred
A diacylglycerol containing palmitic acid and α-linolenic acid at the sn-1 and sn-3 positions, respectively; has been found in the peel and pulp of unripe Dwarf Cavendish bananas
Catalog No. 50-293-4200 Supplier Cayman Chemical Supplier No. 2682810 MG
A diacylglycerol containing palmitic acid and α-linolenic acid at the sn-1 and sn-3 positions, respectively; has been found in the peel and pulp of unripe Dwarf Cavendish bananas