Learn More
Cayman Chemical 1-Palmitoyl-3-Linolenoyl-rac-glycerol, Purity: ≥98%, CAS: 126281-90-1, 50 mg
SDP
Catalog No. 502934204
Encompass_Preferred
A diacylglycerol containing palmitic acid and α-linolenic acid at the sn-1 and sn-3 positions, respectively; has been found in the peel and pulp of unripe Dwarf Cavendish bananas
Catalog No. 50-293-4204 Supplier Cayman Chemical Supplier No. 2682850 MG
A diacylglycerol containing palmitic acid and α-linolenic acid at the sn-1 and sn-3 positions, respectively; has been found in the peel and pulp of unripe Dwarf Cavendish bananas