Learn More
Cayman Chemical 1-Palmitoyl-3-Stearoyl-rac-glycerol, Purity: ≥98%, CAS: 17708-08-6, 10 mg
SDP
Catalog No. 502934538
Encompass_Preferred
A diacylglycerol; has been found in palm-based diacylglycerols, as well as in wheat bran and brewer’s grain extracts
Catalog No. 50-293-4538 Supplier Cayman Chemical Supplier No. 2687410 MG
A diacylglycerol; has been found in palm-based diacylglycerols, as well as in wheat bran and brewer’s grain extracts