Learn More
Cayman Chemical 1PalmItoyld92hydroxysng 100ug
SDP
Catalog No. 502949266
Encompass_Preferred
An internal standard for the quantification of 1-palmitoyl LPA acid by GC- or LC-MS
Catalog No. 50-294-9266 Supplier Cayman Chemical Supplier No. 33479100 UG
An internal standard for the quantification of 1-palmitoyl LPA acid by GC- or LC-MS