Learn More
Cayman Chemical 1PalmItoyld92PalmItoylsn 100ug
SDP
Catalog No. 502939647
Encompass_Preferred
An internal standard for the quantification of 1,2-dipalmitoyl-sn-glycero-3-PC by GC- or LC-MS
Catalog No. 50-293-9647 Supplier Cayman Chemical Supplier No. 28154100 UG
An internal standard for the quantification of 1,2-dipalmitoyl-sn-glycero-3-PC by GC- or LC-MS