Learn More
Cayman Chemical 1PalmItoyld92PalmItoylsn 100ug
SDP
Supplier:  Cayman Chemical 28953100 UG
Encompass_Preferred
An internal standard for the quantification of 1,2-dipalmitoyl-sn-glycero-3-PG
Catalog No. 50-294-0913