Learn More
Cayman Chemical 1Pentadecnoylracglycerol 100mg
SDP
Catalog No. 502939407
Encompass_Preferred
A monoacylglycerol; has been found in wheat bran extracts; levels are increased in a HepaRG cell-based model of hepatic steatosis induced by BSA-conjugated palmitate
Catalog No. 50-293-9407 Supplier Cayman Chemical Supplier No. 26959100 MG
A monoacylglycerol; has been found in wheat bran extracts; levels are increased in a HepaRG cell-based model of hepatic steatosis induced by BSA-conjugated palmitate