Learn More
Cayman Chemical 1-Pyrenesulfonic Acid (sodium salt), Purity: ≥90%, CAS: 59323-54-5, 500 mg
SDP
Catalog No. 502939448
Encompass_Preferred
A fluorescent probe; abs/em = 346/376 nm, respectively; fluorescence can be quenched by ferric ions and the nitroaromatic compounds and explosive components 2,4-DNT and TNT in aqueous solutions; fluorescence quenching has been used in study of reverse micelles, including intermicellar migration of reactants and in the estimation of the number of micelles per cluster; has also been used as a surfactant and dopant in the synthesis of polypyrrole micro- and nanowires
Catalog No. 50-293-9448 Supplier Cayman Chemical Supplier No. 27983500 MG
A fluorescent probe; abs/em = 346/376 nm, respectively; fluorescence can be quenched by ferric ions and the nitroaromatic compounds and explosive components 2,4-DNT and TNT in aqueous solutions; fluorescence quenching has been used in study of reverse micelles, including intermicellar migration of reactants and in the estimation of the number of micelles per cluster; has also been used as a surfactant and dopant in the synthesis of polypyrrole micro- and nanowires