Learn More
Starna Cells Inc 1Q1 STD RECT QUARTZ PATH LGTH1
SDP
Catalog No. NC9744767
Encompass_Preferred
Macro/Standard Rectangular, w/lid, Spectrosil quartz, 10W Internal, 3.5L x 12.5W x 45H, 0.400mL, Open top, supplied with non-sealing PTFE cover, Two polished windows, Walls polished internally, Fine ground externally
Catalog No. NC9744767 Supplier Starna Cells Inc Supplier No. 1Q1
Macro/Standard Rectangular, w/lid, Spectrosil quartz, 10W Internal, 3.5L x 12.5W x 45H, 0.400mL, Open top, supplied with non-sealing PTFE cover, Two polished windows, Walls polished internally, Fine ground externally