Learn More
Tecan Range B Dye for QC Kit
Supplier:  Tecan 30062841
Encompass_Preferred
96-well volume range: 10-49. 99 L, 384-well volume range: 2. 5-9. 99 L. 1 bottle/sales unit (110 mL). The Tecan QC Kit is a quality control tool for pipetting volume verification of your liquid handling platform, ensuring full regulatory compliance.
Catalog No. NC1321973