Learn More
Ace Glass, Inc. 1qt high temperature silicone based bath oil
SDP
Catalog No. 509559121
Encompass
An extremely stable, medium viscosity, silicone oil. Temperature range: High Temp. maximum 230 degree C. Oil shows no discoloration after 40 hours at maximum temperature.
Catalog No. 50-955-9121 Supplier Ace Glass, Inc. Supplier No. 1411512
An extremely stable, medium viscosity, silicone oil. Temperature range: High Temp. maximum 230 degree C. Oil shows no discoloration after 40 hours at maximum temperature.