Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-(-)-1-Amino-2-indanol 98.0+%, TCI America™
SDP

Catalog No. A23061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23061G 1 g
A23065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23061G Supplier TCI America Supplier No. A23061G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 163061-73-2
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD08275447
InChI Key LOPKSXMQWBYUOI-RKDXNWHRSA-N
Synonym (1R,2R)-(-)-1-Amino-2-hydroxyindan
PubChem CID 6931154
IUPAC Name (1R,2R)-1-amino-2,3-dihydro-1H-inden-2-ol
SMILES C1C(C(C2=CC=CC=C21)N)O

Specifications

Melting Point 144°C
Quantity 1 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-(-)-1-Amino-2-indanol
EINECSNumber (4)-1717
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.