Learn More
Medchemexpress LLC (1R,2R)-1-Aminoindan-2-ol | 163061-73-2 | 149.19 | 500 MG
SDP
Catalog No. 5000445832
Encompass_Preferred
(1R,2R)-1-Aminoindan-2-ol is an isomer of (1S,2R)-1-Aminoindan-2-ol. It can be utilized as a biological material or organic compound for life science research. This product can also serve as a starting material for preparing an indene-based chiral auxiliary used in aldol reactions, and in the synthesis of oxazoline-alcohol ligands.
  • Utilized as a biological material
  • Organic compound for life science research
  • Starting material for preparing an indene-based chiral auxiliary used in aldol reactions
  • Starting material in the synthesis of oxazoline-alcohol ligands
Catalog No. 50-004-45832 Supplier Medchemexpress LLC Supplier No. HYW002530A500MG
(1R,2R)-1-Aminoindan-2-ol is an isomer of (1S,2R)-1-Aminoindan-2-ol. It can be utilized as a biological material or organic compound for life science research. This product can also serve as a starting material for preparing an indene-based chiral auxiliary used in aldol reactions, and in the synthesis of oxazoline-alcohol ligands.
  • Utilized as a biological material
  • Organic compound for life science research
  • Starting material for preparing an indene-based chiral auxiliary used in aldol reactions
  • Starting material in the synthesis of oxazoline-alcohol ligands