Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R,2R)-2-pcca (hydrochloride) | 1609563-71-4 | 98.1% | 528.56 | C30H39Cl2N3O | 1 ML
SDP

Catalog No. 5000215562
Encompass_Preferred

(1R,2R)-2-PCCA hydrochloride is the (1R,2R) diastereomer of 2-PCCA and is used as a pharmacological agonist to study GPR88 signaling in biochemical and cell-based assays. It is available as a solid and as a pre-prepared 10 mM solution in DMSO for convenient use in experiments.

  • Potent GPR88 receptor agonist with low-nanomolar activity in cell-free assays.
  • Documented cell-based activity and functional inhibition of GPR88-mediated cAMP.
  • Molecular weight 528.56 and formula C30H39Cl2N3O for precise dosing calculations.
  • High purity (98.08%) with enantiomeric excess ~99% for stereochemical consistency.
  • Available as solid quantities and as 10 mM solution in DMSO for immediate use.
  • Storage recommendations provided for solids and solutions to preserve stability.

Catalog No. 50-002-15562 Supplier Medchemexpress LLC Supplier No. HY100013A110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1R,2R)-2-PCCA hydrochloride is the (1R,2R) diastereomer of 2-PCCA and is used as a pharmacological agonist to study GPR88 signaling in biochemical and cell-based assays. It is available as a solid and as a pre-prepared 10 mM solution in DMSO for convenient use in experiments.

  • Potent GPR88 receptor agonist with low-nanomolar activity in cell-free assays.
  • Documented cell-based activity and functional inhibition of GPR88-mediated cAMP.
  • Molecular weight 528.56 and formula C30H39Cl2N3O for precise dosing calculations.
  • High purity (98.08%) with enantiomeric excess ~99% for stereochemical consistency.
  • Available as solid quantities and as 10 mM solution in DMSO for immediate use.
  • Storage recommendations provided for solids and solutions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.