Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R,2R)-2-PCCA hydrochloride | 1609563-71-4 | 98.1% | 528.56 | C30H39Cl2N3O | 25 MG
SDP

Catalog No. 5000215569
Encompass_Preferred

(1R,2R)-2-PCCA hydrochloride is the (1R,2R) diastereomer of 2-PCCA, a research-stage small molecule that functions as a GPR88 receptor agonist. It is used in in vitro pharmacology to characterize GPR88 activity and signaling and is supplied as a hydrochloride salt for laboratory assays.

  • Potent GPR88 receptor agonist (EC50 = 3 nM in cell-free assay).
  • Shows cellular activity with EC50 = 603 nM in cell-based assays.
  • Inhibits GPR88-mediated cAMP production (EC50 = 56 nM) in HEK293 GPR88/GloSensor-22F assays.
  • Molecular weight 528.56 and molecular formula C30H39Cl2N3O.
  • High enantiomeric purity (98.1% ee) and reported purity 99.0%.
  • Available in multiple vial sizes for laboratory research use.

Catalog No. 50-002-15569 Supplier Medchemexpress LLC Supplier No. HY100013A125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1R,2R)-2-PCCA hydrochloride is the (1R,2R) diastereomer of 2-PCCA, a research-stage small molecule that functions as a GPR88 receptor agonist. It is used in in vitro pharmacology to characterize GPR88 activity and signaling and is supplied as a hydrochloride salt for laboratory assays.

  • Potent GPR88 receptor agonist (EC50 = 3 nM in cell-free assay).
  • Shows cellular activity with EC50 = 603 nM in cell-based assays.
  • Inhibits GPR88-mediated cAMP production (EC50 = 56 nM) in HEK293 GPR88/GloSensor-22F assays.
  • Molecular weight 528.56 and molecular formula C30H39Cl2N3O.
  • High enantiomeric purity (98.1% ee) and reported purity 99.0%.
  • Available in multiple vial sizes for laboratory research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.