Learn More
Strem, An Ascensus Company CAS# 144222-34-4. 2g. (1R,2R)-(-)-N-(4-toluenesulfonyl)-1,2-diphenylethylenediamine, 98% (R,R)-TsDPEN. MFCD02093428
SDP
Catalog No. 5090109142
Encompass
CAS# 144222-34-4. 2g. (1R,2R)-(-)-N-(4-toluenesulfonyl)-1,2-diphenylethylenediamine, 98% (R,R)-TsDPEN. MFCD02093428. Molecular Weight: 366.48. Molecular Formula: C21H22N2O2S. Color/form: white xtl. Strem# 07-2371. http://www.strem.com/catalog/v/07-2371/
Catalog No. 50-901-09142 Supplier Strem, An Ascensus Company Supplier No. 0723712G
CAS# 144222-34-4. 2g. (1R,2R)-(-)-N-(4-toluenesulfonyl)-1,2-diphenylethylenediamine, 98% (R,R)-TsDPEN. MFCD02093428. Molecular Weight: 366.48. Molecular Formula: C21H22N2O2S. Color/form: white xtl. Strem# 07-2371. http://www.strem.com/catalog/v/07-2371/