Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-N,N'-Bis[3-oxo-2-(2,4,6-trimethylbenzoyl)butylidene]-1,2-diphenylethylenediaminato Cobalt(II), TCI America™
SDP

Catalog No. B1844100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1844100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1844100MG Supplier TCI America Supplier No. B1844100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 212250-92-5
Molecular Formula C42H44CoN2O4
Molecular Weight (g/mol) 699.76
MDL Number MFCD01321131
InChI Key KKCIKZXUKHOLHL-UHFFFAOYNA-N
Synonym N,N′C-Bis[2-(mesitoyl)-3-oxobutylidene]-(1R,2R)-1,2-diphenylethylenediaminato Cobalt(II), (R)-MPAC
PubChem CID 71773594
IUPAC Name 2-{[(2-{[3-oxo-2-(2,4,6-trimethylbenzoyl)but-1-en-1-yl]amino}-1,2-diphenylethyl)amino]methylidene}-1-(2,4,6-trimethylphenyl)butane-1,3-dione cobalt
SMILES [Co].CC(=O)C(=CNC(C(NC=C(C(C)=O)C(=O)C1=C(C)C=C(C)C=C1C)C1=CC=CC=C1)C1=CC=CC=C1)C(=O)C1=C(C)C=C(C)C=C1C

Specifications

Quantity 100 mg
Formula Weight 697.74
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2R)-N,N′-Bis[3-oxo-2-(2,4,6-trimethylbenzoyl)butylidene]-1,2-diphenylethylenediaminato Cob
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.