Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1R,2R)-(+)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine, 99%

Catalog No. AC294662500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294662500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC294662500 Supplier Thermo Scientific Chemicals Supplier No. 294662500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 118628-68-5 | C16H20N2 | 240.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118628-68-5
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00216955
InChI Key BTHYVQUNQHWNLS-HZPDHXFCSA-N
Synonym 1r,2r-n1,n2-dimethyl-1,2-diphenylethane-1,2-diamine, 1r,2r-n,n'-dimethyl-1,2-diphenyl-1,2-ethanediamine, methyl 1r,2r-2-methylamino-1,2-diphenylethyl amine, 1r,2r-n,n'-dimethyl-1,2-diphenyl-1,2-ethylenediamine, 1r,2r-n,n'-dimethyl-1,2-diphenylethylenediamine, 1r,2r-n,n/'-dimethyl-1,2-diphenylethylenediamine, 1r,2r-n,n'-dimethyl-1,2-diphenylethane-1,2-diamine, 1r,2r-n,n'-dimethyl-1,2-diphenyl-ethane-1,2-diamine, 1r,2r-n∼1∼,n∼2∼-dimethyl-1,2-diphenylethane-1,2-diamine
PubChem CID 2724997
IUPAC Name (1R,2R)-N,N'-dimethyl-1,2-diphenylethane-1,2-diamine
SMILES CNC(C1=CC=CC=C1)C(C2=CC=CC=C2)NC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 48.0°C to 51.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 250 mg
Fieser 16,159; 16,159; 16,153; 16,153; 17,127
Specific Rotation Condition + 49.50 (20.00°C c=2,MeOH)
Specific Rotation + 49.50
Formula Weight 240.35
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material (1R,2R)-(+)-N,N′-Dimethyl-1,2-diphenyl-1,2-ethane diamine, 99%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.