Learn More
Medchemexpress LLC (1R,2R)-N1,N2-Dibenzylcyclohexane-1,2-diamine | 143443-23-6 | MFCD29919494 | 97.7% | 294.44 g·mol⁻¹ | C20H26N2 | 250 MG
SDP
Catalog No. 5000303579
Encompass_Preferred
(1R,2R)-N1,N2-Dibenzylcyclohexane-1,2-diamine is a chiral diamine used primarily as a synthetic intermediate in the preparation of pharmaceutical and fine chemical compounds. It is intended for research and development applications such as route scouting, asymmetric synthesis, and medicinal chemistry studies.
  • Chiral diamine with defined (1R,2R) stereochemistry.
  • Drug intermediate for synthesis of active pharmaceutical compounds.
  • High purity (≈97.7%) suitable for analytical and synthetic use.
  • Molecular formula C20H26N2 and molecular weight 294.44 g·mol⁻¹.
  • Available in multiple small-scale pack sizes for research use.
Catalog No. 50-003-03579 Supplier Medchemexpress LLC Supplier No. 250MGHYW037146
(1R,2R)-N1,N2-Dibenzylcyclohexane-1,2-diamine is a chiral diamine used primarily as a synthetic intermediate in the preparation of pharmaceutical and fine chemical compounds. It is intended for research and development applications such as route scouting, asymmetric synthesis, and medicinal chemistry studies.
  • Chiral diamine with defined (1R,2R) stereochemistry.
  • Drug intermediate for synthesis of active pharmaceutical compounds.
  • High purity (≈97.7%) suitable for analytical and synthetic use.
  • Molecular formula C20H26N2 and molecular weight 294.44 g·mol⁻¹.
  • Available in multiple small-scale pack sizes for research use.