Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R,2R)-N1,N2-dibenzylcyclohexane-1,2-diamine | 143443-23-6 | MFCD29919494 | 97.7% | 294.44 g/mol | C20H26N2 | 5 G
SDP

Catalog No. 5000299710
Encompass_Preferred

(1R,2R)-N1,N2-dibenzylcyclohexane-1,2-diamine is a chiral N,N'-dibenzyl-substituted 1,2-cyclohexanediamine used as a drug intermediate in medicinal chemistry. It is supplied with analytical documentation to support research and synthetic applications.

  • Chiral diamine useful as a drug intermediate for synthesis of active compounds.
  • CAS 143443-23-6.
  • Chemical formula C20H26N2.
  • Molecular weight 294.44 g/mol.
  • Purity 97.7% (HPLC).
  • Available in small pack sizes for research use (100 mg-5 g).
  • Certificate of analysis and safety data sheet available.

Catalog No. 50-002-99710 Supplier Medchemexpress LLC Supplier No. HYW0371465G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1R,2R)-N1,N2-dibenzylcyclohexane-1,2-diamine is a chiral N,N'-dibenzyl-substituted 1,2-cyclohexanediamine used as a drug intermediate in medicinal chemistry. It is supplied with analytical documentation to support research and synthetic applications.

  • Chiral diamine useful as a drug intermediate for synthesis of active compounds.
  • CAS 143443-23-6.
  • Chemical formula C20H26N2.
  • Molecular weight 294.44 g/mol.
  • Purity 97.7% (HPLC).
  • Available in small pack sizes for research use (100 mg-5 g).
  • Certificate of analysis and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.