Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1R,2R)-N1,N2-dibenzylcyclohexane-1,2-diamine | 143443-23-6 | MFCD29919494 | 97.7% | 294.44 g/mol | C20H26N2 | 500 MG
SDP

Catalog No. 5000301193
Encompass_Preferred

(1R,2R)-N1,N2-dibenzylcyclohexane-1,2-diamine is a chiral diamine intermediate used in organic synthesis and medicinal chemistry. The (1R,2R) stereochemistry and dibenzyl substitution make it useful as a building block for asymmetric synthesis and for preparing biologically active compounds.

  • Chiral diamine with defined (1R,2R) stereochemistry.
  • High purity: 97.7% by HPLC (as provided).
  • Chemical formula C20H26N2 and molecular weight 294.44 g/mol.
  • Suitable as a synthetic intermediate for drug discovery and ligand synthesis.
  • Storage recommendations: 4°C, protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-01193 Supplier Medchemexpress LLC Supplier No. HYW037146500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1R,2R)-N1,N2-dibenzylcyclohexane-1,2-diamine is a chiral diamine intermediate used in organic synthesis and medicinal chemistry. The (1R,2R) stereochemistry and dibenzyl substitution make it useful as a building block for asymmetric synthesis and for preparing biologically active compounds.

  • Chiral diamine with defined (1R,2R) stereochemistry.
  • High purity: 97.7% by HPLC (as provided).
  • Chemical formula C20H26N2 and molecular weight 294.44 g/mol.
  • Suitable as a synthetic intermediate for drug discovery and ligand synthesis.
  • Storage recommendations: 4°C, protect from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.