Learn More
Medchemexpress LLC (1R,2S)-1-tert-Butoxycarbonylamino-2-vinylcyclopropanecarboxylic acid | 159622-10-3 | 99.88% | 227.26 | 10 G
SDP
Catalog No. 5000401220
Encompass_Preferred
(1R,2S)-1-tert-Butoxycarbonylamino-2-vinylcyclopropanecarboxylic acid is a chemical compound suitable for laboratory research. It appears as a white to off-white solid with a high purity of 99.88%. It is used for manufacturing substances and various laboratory applications.
  • High purity (99.88%)
  • White to off-white solid appearance
  • Specific storage conditions for powder (-20°C for 3 years, 4°C for 2 years)
  • Specific storage conditions when in solvent (-80°C for 6 months, -20°C for 1 month)
  • Identified for laboratory chemicals and substance manufacturing
Catalog No. 50-004-01220 Supplier Medchemexpress LLC Supplier No. HY3048610G
(1R,2S)-1-tert-Butoxycarbonylamino-2-vinylcyclopropanecarboxylic acid is a chemical compound suitable for laboratory research. It appears as a white to off-white solid with a high purity of 99.88%. It is used for manufacturing substances and various laboratory applications.
  • High purity (99.88%)
  • White to off-white solid appearance
  • Specific storage conditions for powder (-20°C for 3 years, 4°C for 2 years)
  • Specific storage conditions when in solvent (-80°C for 6 months, -20°C for 1 month)
  • Identified for laboratory chemicals and substance manufacturing